Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027840.3 | NP_082116.1 | 2 - 3 | 282 | 72 | |
GenBank mRNA | AF395844.1 | - | 1 - 2 | 121 | 72 | |
AK018632.1 | - | 2 - 3 | 284 | 72 | ||
AK042068.1 | - | 2 - 3 | 257 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |