Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080643.2 | NP_542374.2 | 5 - 6 | 865 | 63 | |
GenBank mRNA | AF451978.1 | - | 4 - 5 | 584 | 63 | |
AK129297.1 | - | 5 - 6 | 903 | 63 | ||
BC053083.1 | - | 5 - 6 | 865 | 5704642 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |