Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100009605.mm.1 |
NM_001102611.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009605.mm.1 |
NM_001102611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009605.mm.1 |
TC1100001066.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100009604.mm.1 |
NM_001102611.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009604.mm.1 |
NM_001102611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009604.mm.1 |
TC1100001066.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100009606.mm.1 |
NM_001102611.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009606.mm.1 |
NM_001102611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009606.mm.1 |
TC1100001066.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100009607.mm.1 |
NM_001102611.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009607.mm.1 |
NM_001102611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009607.mm.1 |
TC1100001066.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100009600.mm.1 |
NM_001102611.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009600.mm.1 |
NM_001102611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009600.mm.1 |
TC1100001066.mm.1 |
MTA TC |