Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011631.1 | NP_035761.1 | 5 - 6 | 822 | 144 | |
GenBank mRNA | AK031918.1 | - | 1 - 2 | 174 | 144 | |
AK088148.1 | - | 3 - 4 | 525 | 144 | ||
AK146885.1 | - | 5 - 6 | 888 | 144 | ||
AK150811.1 | - | 5 - 6 | 842 | 144 | ||
AK151413.1 | - | 5 - 6 | 839 | 144 | ||
AK153550.1 | - | 5 - 6 | 842 | 144 | ||
AK159974.1 | - | 5 - 6 | 847 | 144 | ||
AK160093.1 | - | 5 - 6 | 841 | 144 | ||
AK160827.1 | - | 262 | 144 | |||
AK165975.1 | - | 5 - 6 | 842 | 144 | ||
AK168952.1 | - | 5 - 6 | 842 | 144 | ||
AK169090.1 | - | 5 - 6 | 842 | 144 | ||
BC010445.1 | - | 5 - 6 | 754 | 3587178 | 144 | |
BC011439.1 | - | 5 - 6 | 828 | 3707817 | 144 | |
J03297.1 | - | 5 - 6 | 822 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |