Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC151125.1 | - | 2 - 3 | 377 | 9088038 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |