Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010578.2 | NP_034708.1 | 9 - 10 | 1431 | 103 | |
GenBank mRNA | AK088729.1 | - | 9 - 10 | 1178 | 103 | |
AK135521.1 | - | 8 - 9 | 1107 | 103 | ||
AK141253.1 | - | 4 - 5 | 358 | 103 | ||
AK159689.1 | - | 9 - 10 | 1273 | 103 | ||
AK167682.1 | - | 9 - 10 | 1329 | 103 | ||
BC050906.1 | - | 9 - 10 | 1304 | 5721348 | 103 | |
Y00769.1 | - | 9 - 10 | 1219 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |