Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197995.2 | NP_932112.2 | 3 - 4 | 408 | 100 | |
GenBank mRNA | AK038349.1 | - | 3 - 4 | 280 | 100 | |
AK140077.1 | - | 2 - 3 | 412 | 100 | ||
BC139260.1 | - | 3 - 4 | 279 | 8862282 | 100 | |
BC139262.1 | - | 3 - 4 | 279 | 8862284 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |