Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029746.3 | NP_084022.2 | 1 - 2 | 183 | 62 | |
GenBank mRNA | AK012236.1 | - | 1 - 2 | 117 | 62 | |
AK089920.1 | - | 1 - 2 | 167 | 62 | ||
AK157348.1 | - | 1 - 2 | 145 | 62 | ||
AK169298.1 | - | 1 - 2 | 153 | 62 | ||
AK171726.1 | - | 1 - 2 | 102 | 62 | ||
BC011435.1 | - | 1 - 2 | 123 | 4010102 | 62 | |
CJ093818.1 | - | 1 - 2 | 183 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |