Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053261.2 | NP_444491.1 | 4 - 5 | 572 | 68 | |
GenBank mRNA | AF353730.1 | - | 4 - 5 | 571 | 68 | |
AK154489.1 | - | 4 - 5 | 610 | 68 | ||
AK154845.1 | - | 4 - 5 | 543 | 68 | ||
AK170962.1 | - | 4 - 5 | 610 | 68 | ||
AK171626.1 | - | 4 - 5 | 545 | 68 | ||
BC011093.1 | - | 4 - 5 | 547 | 4012163 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |