Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100018954.mm.1 |
NM_001001883.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018954.mm.1 |
NM_001001883.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018954.mm.1 |
NM_172655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018954.mm.1 |
NM_172655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018954.mm.1 |
TC0100002338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018951.mm.1 |
NM_001001883.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018951.mm.1 |
NM_001001883.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018951.mm.1 |
NM_172655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018951.mm.1 |
NM_172655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018951.mm.1 |
TC0100002338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018950.mm.1 |
NM_001001883.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018950.mm.1 |
NM_001001883.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018950.mm.1 |
NM_172655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018950.mm.1 |
NM_172655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018950.mm.1 |
TC0100002338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018952.mm.1 |
NM_001001883.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018952.mm.1 |
NM_001001883.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018952.mm.1 |
NM_172655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018952.mm.1 |
NM_172655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018952.mm.1 |
TC0100002338.mm.1 |
MTA TC |