Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100449.1 | NP_001093919.1 | 2 - 3 | 835 | 83 | |
GenBank mRNA | AK012135.1 | - | 2 - 3 | 837 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |