Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009509.2 | NP_033535.2 | 2 - 3 | 140 | 62 | |
GenBank mRNA | AI663456.1 | - | 2 - 3 | 132 | 62 | |
BB871941.1 | - | 2 - 3 | 141 | 62 | ||
BC015267.1 | - | 2 - 3 | 116 | 4236751 | 62 | |
BC117875.2 | - | 2 - 3 | 105 | 40095718 | 62 | |
BC145671.1 | - | 2 - 3 | 107 | 40130974 | 62 | |
M98454.1 | - | 2 - 3 | 130 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |