Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arntl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289679.1 | NP_001276608.1 | 2 - 3 | 152 | 89 | |
NM_001289680.1 | NP_001276609.1 | 2 - 3 | 152 | 89 | ||
NM_001289681.1 | NP_001276610.1 | 2 - 3 | 152 | 89 | ||
NM_172309.2 | NP_758513.1 | 2 - 3 | 152 | 89 | ||
GenBank mRNA | AY005163.1 | - | 2 - 3 | 152 | 89 | |
AY014836.1 | - | 2 - 3 | 152 | 89 | ||
BC108965.2 | - | 2 - 3 | 152 | 40047159 | 89 | |
BC108966.1 | - | 2 - 3 | 152 | 89 | ||
DQ322241.1 | - | 2 - 3 | 152 | 89 | ||
DQ355143.1 | - | 2 - 3 | 152 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |