Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam117b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037725.3 | NP_001032814.1 | 2 - 3 | 893 | 88 | |
GenBank mRNA | AK154812.1 | - | 2 - 3 | 895 | 88 | |
BC080299.1 | - | 2 - 3 | 288 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |