Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008419.2 | NP_001008419.1 | 11 - 12 | 1159 | 75 | |
GenBank mRNA | AY187018.2 | - | 11 - 12 | 1159 | 75 | |
AY665589.1 | - | 11 - 12 | 1066 | 75 | ||
BC150826.1 | - | 11 - 12 | 1086 | 9087737 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |