Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011770.4 | NP_035900.2 | 2 - 3 | 322 | 89 | |
GenBank mRNA | AF044257.1 | - | 2 - 3 | 217 | 89 | |
AK047925.1 | - | 2 - 3 | 184 | 89 | ||
BC050049.1 | - | 2 - 3 | 155 | 89 | ||
BC138606.1 | - | 2 - 3 | 162 | 8861626 | 89 | |
BC138608.1 | - | 2 - 3 | 162 | 8861628 | 89 | |
CJ161925.1 | - | 2 - 3 | 322 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |