Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039554.3 | NP_001034643.1 | 3 - 4 | 815 | 111 | |
GenBank mRNA | AK031675.1 | - | 3 - 4 | 815 | 111 | |
BC023373.1 | - | 3 - 4 | 802 | 5043159 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |