Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009533.2 | NP_033559.2 | 9 - 10 | 1175 | 88 | |
GenBank mRNA | AF166486.1 | - | 9 - 10 | 1070 | 88 | |
AK081633.1 | - | 9 - 10 | 1176 | 88 | ||
AK089944.1 | - | 9 - 10 | 1172 | 88 | ||
AK165470.1 | - | 9 - 10 | 1143 | 88 | ||
AK167312.1 | - | 9 - 10 | 1064 | 88 | ||
AK168913.1 | - | 9 - 10 | 1064 | 88 | ||
AK169838.1 | - | 9 - 10 | 1064 | 88 | ||
BC029218.1 | - | 9 - 10 | 1047 | 88 | ||
BC051660.1 | - | 9 - 10 | 1115 | 30008031 | 88 | |
X66323.1 | - | 9 - 10 | 1076 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |