Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Far2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178797.3 | NP_848912.1 | 5 - 6 | 861 | 70 | |
GenBank mRNA | AK038584.1 | - | 5 - 6 | 814 | 70 | |
AK043388.1 | - | 5 - 6 | 861 | 70 | ||
AK043589.1 | - | 5 - 6 | 861 | 70 | ||
AK137298.1 | - | 5 - 6 | 861 | 70 | ||
AK161201.1 | - | 5 - 6 | 828 | 70 | ||
BC055759.1 | - | 5 - 6 | 839 | 6809131 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |