Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025613.1 | NP_001020784.1 | 11 - 12 | 1655 | 69 | |
NM_001025614.1 | NP_001020785.1 | 10 - 11 | 1597 | 69 | ||
GenBank mRNA | AK044701.1 | - | 11 - 12 | 1656 | 69 | |
BC009124.1 | - | 5 - 6 | 564 | 69 | ||
BC037040.1 | - | 13 - 14 | 1641 | 69 | ||
BC141397.1 | - | 10 - 11 | 1487 | 9056010 | 69 | |
BC141398.1 | - | 10 - 11 | 1487 | 9056011 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |