Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290549.1 | NP_001277478.1 | 12 - 13 | 2255 | 120 | |
NM_001290551.1 | NP_001277480.1 | 11 - 12 | 2102 | 120 | ||
NM_013690.3 | NP_038718.2 | 12 - 13 | 2255 | 120 | ||
GenBank mRNA | BC050824.1 | - | 11 - 12 | 2096 | 6333576 | 120 |
D13738.1 | - | 12 - 13 | 2257 | 120 | ||
S67051.1 | - | 13 - 14 | 1918 | 120 | ||
X67553.1 | - | 12 - 13 | 2026 | 120 | ||
X71426.1 | - | 13 - 14 | 1989 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |