Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290549.1 | NP_001277478.1 | 9 - 10 | 1675 | 126 | |
NM_001290551.1 | NP_001277480.1 | 8 - 9 | 1522 | 126 | ||
NM_013690.3 | NP_038718.2 | 9 - 10 | 1675 | 126 | ||
GenBank mRNA | BC050824.1 | - | 8 - 9 | 1516 | 6333576 | 126 |
D13738.1 | - | 9 - 10 | 1677 | 126 | ||
S67051.1 | - | 10 - 11 | 1338 | 126 | ||
X67553.1 | - | 9 - 10 | 1446 | 126 | ||
X71426.1 | - | 10 - 11 | 1409 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |