Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026539.2 | NP_080815.1 | 4 - 5 | 463 | 97 | |
GenBank mRNA | AK011061.1 | - | 4 - 5 | 477 | 97 | |
AK014473.1 | - | 4 - 5 | 465 | 97 | ||
AK165656.1 | - | 4 - 5 | 487 | 97 | ||
BC019617.2 | - | 4 - 5 | 462 | 97 | ||
BC052385.1 | - | 4 - 5 | 447 | 5703859 | 97 | |
BC057567.1 | - | 4 - 5 | 476 | 6415898 | 97 | |
BC062966.1 | - | 4 - 5 | 476 | 6415898 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |