Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008718.2 | NP_032744.1 | 8 - 9 | 1154 | 62 | |
GenBank mRNA | BC132113.1 | - | 7 - 8 | 989 | 40130390 | 62 |
BC137863.1 | - | 7 - 8 | 989 | 8860882 | 62 | |
U77967.1 | - | 7 - 8 | 1022 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |