Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001488.3 | NP_001001488.2 | 9 - 10 | 1105 | 105 | |
GenBank mRNA | AK036959.1 | - | 9 - 10 | 1026 | 105 | |
AK157316.1 | - | 9 - 10 | 1373 | 105 | ||
AY506548.1 | - | 9 - 10 | 834 | 105 | ||
BC117946.2 | - | 9 - 10 | 832 | 40098035 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |