Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177407.4 | NP_803126.1 | 12 - 13 | 1091 | 77 | |
GenBank mRNA | AK083245.1 | - | 12 - 13 | 1092 | 77 | |
AK122410.2 | - | 12 - 13 | 1087 | 77 | ||
BC031745.1 | - | 12 - 13 | 1072 | 4506005 | 77 | |
X14836.1 | - | 12 - 13 | 1075 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |