Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134134.2 | NP_598895.2 | 4 - 5 | 1280 | 60 | |
NM_178277.1 | NP_840061.1 | 4 - 5 | 1279 | 60 | ||
GenBank mRNA | AB045322.1 | - | 4 - 5 | 1280 | 60 | |
AK041866.1 | - | 4 - 5 | 1258 | 60 | ||
AK129085.1 | - | 1 - 2 | 187 | 60 | ||
AK144993.1 | - | 4 - 5 | 1264 | 60 | ||
BC058089.1 | - | 2 - 3 | 577 | 60 | ||
BC079580.1 | - | 4 - 5 | 1227 | 5702202 | 60 | |
BU611563.1 | - | 1 - 2 | 391 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |