Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400016822.mm.1 |
NM_001162977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016822.mm.1 |
NM_001162977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016822.mm.1 |
TC0400002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016820.mm.1 |
NM_001162977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016820.mm.1 |
NM_001162977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016820.mm.1 |
TC0400002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016823.mm.1 |
NM_001162977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016823.mm.1 |
NM_001162977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016823.mm.1 |
TC0400002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016821.mm.1 |
NM_001162977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016821.mm.1 |
NM_001162977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016821.mm.1 |
TC0400002043.mm.1 |
MTA TC |