Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133708.3 | NP_598469.1 | 1 - 2 | 69 | 72 | |
GenBank mRNA | AK007463.1 | - | 1 - 2 | 60 | 72 | |
AK182835.1 | - | 1 - 2 | 29 | 72 | ||
AK189202.1 | - | 1 - 2 | 29 | 72 | ||
BY063474.1 | - | 1 - 2 | 76 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |