Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023732.2 | NP_076221.1 | 17 - 18 | 2514 | 77 | |
GenBank mRNA | AK004605.1 | - | 17 - 18 | 2594 | 77 | |
AK151165.1 | - | 17 - 18 | 2623 | 77 | ||
AK164003.1 | - | 14 - 15 | 3432 | 77 | ||
AK168642.1 | - | 17 - 18 | 2615 | 77 | ||
BC006634.1 | - | 17 - 18 | 2514 | 3710552 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |