Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028355.3 | NP_082631.1 | 3 - 4 | 592 | 105 | |
GenBank mRNA | AK013409.1 | - | 3 - 4 | 329 | 105 | |
AK144734.1 | - | 3 - 4 | 593 | 105 | ||
BC021337.1 | - | 3 - 4 | 347 | 5042118 | 105 | |
DQ141695.1 | - | 3 - 4 | 287 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |