Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gntl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178664.5 | NP_848779.2 | 7 - 8 | 702 | 64 | |
GenBank mRNA | AK031361.1 | - | 7 - 8 | 691 | 64 | |
AK043722.1 | - | 7 - 8 | 705 | 64 | ||
AK045461.1 | - | 6 - 7 | 559 | 64 | ||
AK156318.1 | - | 7 - 8 | 680 | 64 | ||
BC062106.1 | - | 1 - 2 | 561 | 64 | ||
BC088993.1 | - | 7 - 8 | 567 | 6416623 | 64 | |
BC125626.1 | - | 7 - 8 | 675 | 40130141 | 64 | |
BC132193.1 | - | 7 - 8 | 675 | 40130470 | 64 | |
BC145163.1 | - | 7 - 8 | 679 | 64 | ||
BC145165.1 | - | 7 - 8 | 741 | 64 | ||
BC145166.1 | - | 7 - 8 | 675 | 9053717 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |