Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033573.1 | NP_001028745.1 | 2 - 3 | 824 | 94 | |
NM_025883.3 | NP_080159.3 | 2 - 3 | 708 | 94 | ||
GenBank mRNA | AK010296.1 | - | 2 - 3 | 703 | 94 | |
AK011373.1 | - | 2 - 3 | 675 | 94 | ||
AK035172.1 | - | 787 | 94 | |||
AK038177.1 | - | 687 | 94 | |||
AK050388.1 | - | 2 - 3 | 823 | 94 | ||
AK077693.1 | - | 2 - 3 | 662 | 94 | ||
AK077838.1 | - | 673 | 94 | |||
AK139707.1 | - | 2 - 3 | 665 | 94 | ||
AK144128.1 | - | 2 - 3 | 680 | 94 | ||
AK154741.1 | - | 2 - 3 | 686 | 94 | ||
AK157494.1 | - | 2 - 3 | 690 | 94 | ||
AK159304.1 | - | 2 - 3 | 709 | 94 | ||
AK168497.1 | - | 2 - 3 | 703 | 94 | ||
BC033317.1 | - | 2 - 3 | 655 | 4459242 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |