Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053198.3 | NP_444428.3 | 1 - 2 | 130 | 84 | |
GenBank mRNA | AF325263.1 | - | 1 - 2 | 72 | 84 | |
AK148530.1 | - | 1 - 2 | 125 | 84 | ||
BC058211.1 | - | 1 - 2 | 127 | 84 | ||
BC089535.1 | - | 1 - 2 | 112 | 6589805 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |