Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025251.2 | NP_001020422.1 | 2 - 3 | 371 | 58 | |
NM_001025254.2 | NP_001020425.1 | 2 - 3 | 326 | 58 | ||
NM_001025256.2 | NP_001020427.1 | 2 - 3 | 371 | 58 | ||
GenBank mRNA | AK079621.1 | - | 2 - 3 | 327 | 58 | |
AK189263.1 | - | 1 - 2 | 98 | 58 | ||
AK190490.1 | - | 1 - 2 | 98 | 58 | ||
AK191010.1 | - | 1 - 2 | 98 | 58 | ||
AK195158.1 | - | 1 - 2 | 98 | 58 | ||
AK199946.1 | - | 1 - 2 | 98 | 58 | ||
AK203400.1 | - | 1 - 2 | 62 | 58 | ||
BB633793.1 | - | 2 - 3 | 327 | 58 | ||
BC004704.1 | - | 2 - 3 | 288 | 3499588 | 58 | |
CO425201.1 | - | 2 - 3 | 293 | 58 | ||
DQ887821.1 | - | 2 - 3 | 254 | 58 | ||
M15060.1 | - | 2 - 3 | 291 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |