Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4kb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293715.1 | NP_001280644.1 | 10 - 11 | 2525 | 103 | |
NM_001293716.1 | NP_001280645.1 | 8 - 9 | 1543 | 103 | ||
NM_175356.3 | NP_780565.2 | 9 - 10 | 2480 | 103 | ||
GenBank mRNA | AK028974.1 | - | 9 - 10 | 2465 | 103 | |
AK030704.1 | - | 9 - 10 | 2240 | 103 | ||
AK048405.1 | - | 8 - 9 | 1336 | 103 | ||
AK053614.1 | - | 9 - 10 | 2475 | 103 | ||
AK157918.1 | - | 9 - 10 | 2272 | 103 | ||
AK172074.1 | - | 9 - 10 | 2473 | 103 | ||
BC059895.1 | - | 2 - 3 | 227 | 103 | ||
BC079846.1 | - | 9 - 10 | 2480 | 5707855 | 103 | |
BC113176.1 | - | 10 - 11 | 2380 | 40060773 | 103 | |
BC113781.1 | - | 8 - 9 | 1443 | 40060770 | 103 | |
BC138457.1 | - | 9 - 10 | 2383 | 8861477 | 103 | |
BC138458.1 | - | 9 - 10 | 2383 | 8861478 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |