Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800005488.mm.1 |
NM_001289649.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005488.mm.1 |
TC1800000758.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800005489.mm.1 |
NM_001289650.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005489.mm.1 |
TC1800000758.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800005494.mm.1 |
NM_001289648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005494.mm.1 |
NM_001289649.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005494.mm.1 |
NM_001289650.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005494.mm.1 |
NM_029019.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005494.mm.1 |
TC1800000758.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800005495.mm.1 |
NM_001289648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005495.mm.1 |
NM_001289649.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005495.mm.1 |
NM_001289650.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005495.mm.1 |
NM_029019.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005495.mm.1 |
TC1800000758.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800005498.mm.1 |
NM_001289648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005498.mm.1 |
NM_001289649.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005498.mm.1 |
NM_001289650.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005498.mm.1 |
NM_029019.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005498.mm.1 |
TC1800000758.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800005496.mm.1 |
NM_001289648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005496.mm.1 |
NM_001289649.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005496.mm.1 |
NM_001289650.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005496.mm.1 |
NM_029019.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005496.mm.1 |
TC1800000758.mm.1 |
MTA TC |