Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008292.4 | NP_032318.2 | 11 - 12 | 1021 | 86 | |
GenBank mRNA | AK004866.1 | - | 11 - 12 | 1021 | 86 | |
AK088381.1 | - | 11 - 12 | 996 | 86 | ||
AK147182.1 | - | 11 - 12 | 993 | 86 | ||
AK152532.1 | - | 11 - 12 | 916 | 86 | ||
AK161655.1 | - | 11 - 12 | 916 | 86 | ||
AK166801.1 | - | 11 - 12 | 934 | 86 | ||
AK167060.1 | - | 11 - 12 | 971 | 86 | ||
AK169077.1 | - | 11 - 12 | 923 | 86 | ||
BC022175.1 | - | 11 - 12 | 894 | 4209394 | 86 | |
X89998.1 | - | 11 - 12 | 882 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |