Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019514.3 | NP_062387.2 | 16 - 17 | 3078 | 77 | |
NM_207109.2 | NP_996992.1 | 17 - 18 | 3234 | 77 | ||
GenBank mRNA | AF116573.1 | - | 14 - 15 | 2246 | 77 | |
AK041160.1 | - | 18 - 19 | 3138 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |