Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081557.3 | NP_001075026.1 | 9 - 10 | 2754 | 65 | |
GenBank mRNA | AK122383.1 | - | 4 - 5 | 2180 | 65 | |
BC150740.1 | - | 8 - 9 | 2603 | 9087651 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |