Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011155.2 | NP_035285.2 | 3 - 4 | 608 | 67 | |
GenBank mRNA | AF018262.1 | - | 3 - 4 | 579 | 67 | |
BC003744.1 | - | 3 - 4 | 564 | 3590322 | 67 | |
DV653678.1 | - | 3 - 4 | 545 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |