Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800000884.mm.1 |
NM_001163609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000884.mm.1 |
NM_001163609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000884.mm.1 |
TC1800000123.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800000889.mm.1 |
NM_001163609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000889.mm.1 |
NM_001163609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000889.mm.1 |
TC1800000123.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800000887.mm.1 |
NM_001163609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000887.mm.1 |
NM_001163609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000887.mm.1 |
TC1800000123.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800000886.mm.1 |
TC1800000123.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800000885.mm.1 |
NM_001163609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000885.mm.1 |
NM_001163609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000885.mm.1 |
TC1800000123.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800000888.mm.1 |
NM_001163609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000888.mm.1 |
NM_001163609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800000888.mm.1 |
TC1800000123.mm.1 |
MTA TC |