Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008233.3 | NP_001008234.2 | 9 - 10 | 930 | 136 | |
GenBank mRNA | AK048084.1 | - | 7 - 8 | 2909 | 136 | |
BC046544.1 | - | 9 - 10 | 930 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |