Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177796.1 | NP_001171267.1 | 10 - 11 | 1428 | 82 | |
NM_001177797.1 | NP_001171268.1 | 9 - 10 | 1374 | 82 | ||
NM_146102.2 | NP_666214.1 | 11 - 12 | 1480 | 82 | ||
GenBank mRNA | AK048649.1 | - | 11 - 12 | 1134 | 82 | |
AK087449.1 | - | 10 - 11 | 1064 | 82 | ||
AK220430.1 | - | 9 - 10 | 997 | 82 | ||
BC031515.1 | - | 10 - 11 | 1030 | 3711746 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |