Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L1td1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081202.1 | NP_001074671.1 | 1 - 2 | 88 | 50 | |
GenBank mRNA | AB211064.1 | - | 1 - 2 | 94 | 50 | |
AK145814.1 | - | 1 - 2 | 81 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |