Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164583.1 | NP_001158055.1 | 8 - 9 | 1185 | 75 | |
NM_001164584.1 | NP_001158056.1 | 8 - 9 | 1130 | 75 | ||
NM_001164585.1 | NP_001158057.1 | 9 - 10 | 1195 | 75 | ||
NM_198412.2 | NP_940804.1 | 9 - 10 | 1104 | 75 | ||
GenBank mRNA | AK122293.1 | - | 8 - 9 | 1106 | 75 | |
AK147570.1 | - | 9 - 10 | 1067 | 75 | ||
AK147637.1 | - | 9 - 10 | 1067 | 75 | ||
AK163657.1 | - | 8 - 9 | 1004 | 75 | ||
BC035262.1 | - | 3 - 4 | 339 | 75 | ||
BC060734.1 | - | 8 - 9 | 1185 | 6409942 | 75 | |
BC064460.1 | - | 9 - 10 | 1073 | 6821822 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |