Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK155349.1 | - | 7 - 8 | 1077 | 66 | |
BC088987.1 | - | 6 - 7 | 1012 | 66 | ||
BC138848.1 | - | 8 - 9 | 948 | 8861870 | 66 | |
BC138849.1 | - | 8 - 9 | 948 | 8861871 | 66 | |
U10115.1 | - | 7 - 8 | 1034 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |