Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400035019.mm.1 |
TC0400004166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400035019.mm.1 |
NM_029264.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035019.mm.1 |
NM_029264.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035017.mm.1 |
TC0400004166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400035017.mm.1 |
NM_029264.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035017.mm.1 |
NM_029264.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035021.mm.1 |
TC0400004166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400035021.mm.1 |
NM_029264.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035021.mm.1 |
NM_029264.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035020.mm.1 |
TC0400004166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400035020.mm.1 |
NM_029264.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035020.mm.1 |
NM_029264.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035023.mm.1 |
TC0400004166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400035023.mm.1 |
NM_029264.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035023.mm.1 |
NM_029264.2 |
Refseq |