Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pggt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172627.3 | NP_766215.1 | 2 - 3 | 294 | 79 | |
GenBank mRNA | AK028188.1 | - | 2 - 3 | 152 | 79 | |
AK081731.1 | - | 2 - 3 | 295 | 79 | ||
BC141021.1 | - | 2 - 3 | 292 | 9055633 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |