Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048189.4 | NP_001041654.2 | 8 - 9 | 1010 | 65 | |
NM_001284190.1 | NP_001271119.1 | 8 - 9 | 1010 | 65 | ||
NM_030231.3 | NP_084507.1 | 8 - 9 | 1010 | 65 | ||
GenBank mRNA | AK016502.1 | - | 8 - 9 | 975 | 65 | |
DQ867039.1 | - | 8 - 9 | 917 | 65 | ||
DQ867040.1 | - | 8 - 9 | 917 | 65 | ||
FN429928.1 | - | 8 - 9 | 836 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |